Perchloric acid 0.1 M in acetic acid, ren

Leverandør: Thermo Fisher Scientific
Danger

348630010 348630025
ACRO348630010EA 2250 NOK
ACRO348630010 ACRO348630025
Perchloric acid 0.1 M in acetic acid, ren
Perchloric acid
Formel: HClO₄
Tetthet: 1,06 g/cm³
Flash punkt: 40 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011325
CAS-nummer: 7601-90-3
EINECS: 231-512-4
FN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless Solution
Water ≤0.2 % (K.F.)
Strength 0.0998 to 0.1002 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR