Castor olje, meets analytical specification of Ph.Eur.

Leverandør: Thermo Fisher Scientific

Synonymer: Castor oil, Ricinus oil, Oleum Ricini s. Castoris, Oil of Palma Christi

348855000 348850025
ACRO348855000EA 750 NOK
ACRO348855000 ACRO348850025
Castor olje, meets analytical specification of Ph.Eur.
Castor olje
Kokepunkt: 229 °C (760 mmHg)
Smeltepunkt: –10 °C
Tetthet: 0,957 g/cm³
Flash punkt: 229 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00130746
CAS-nummer: 8001-79-4
EINECS: 232-293-8
Merck Indeks: 13,01908

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to slightly yellow Hygroscopic viscous liquid
Water ≤0.3 %
Identification A Passes test
Angle of optical rotation +3.5° to +6.0° (20°C, 589 nm) (neat)
Absorbance ≤0.7 (270 nm, 1 % in EtOH 96%)
Composition of fatty acids ≤1.0 % (Any other fatty acid)
Acid value ≤1.5 mg KOH/g
Hydroxyl value ≥160 mg KOH/g
Peroxide value ≤10.0 meq O2/kg
Unsaponifiable matter ≤0.8 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR