Natriumhydroksid standard solution 0.2 mol/l (0.2 N) in water

Leverandør: Thermo Fisher Scientific
Warning

349685000 349680025
ACRO349685000EA 400 NOK
ACRO349685000 ACRO349680025
Natriumhydroksid standard solution 0.2 mol/l (0.2 N) in water
Natriumhydroksid
Formel: NaOH
Kokepunkt: ∼105 °C (1013 hPa)
Tetthet: 1,01 g/cm³
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless Solution
Strength 0.1996 to 0.2004 N

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR