4-Amino-3-hydroksybenso syre 98%

Leverandør: Thermo Fisher Scientific
Warning

354260050 354260250
ACRO354260050EA 695 NOK
ACRO354260050 ACRO354260250
4-Amino-3-hydroksybenso syre 98%
4-Amino-3-hydroksybenso syre
Formel: C₇H₇NO₃
Smeltepunkt: 211…215 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00017094
CAS-nummer: 2374-03-0

Bestill nå

Spesifikasjon Testresultater

Appearance Yellow-brown to brown Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR