1-Hexadecyl-3-methylimidazolium chloride monohydrate 98%

Leverandør: Thermo Scientific
Warning

Synonymer: 1-Hexadecyl-3-methyl-1H-imidazol-3-ium chloride hydrate

356650100
ACRO356650100EA 2725 NOK
ACRO356650100
1-Hexadecyl-3-methylimidazolium chloride monohydrate 98%
1-Hexadecyl-3-methylimidazolium chloride monohydrate
Formel: C₂₀H₃₉ClN₂·1H₂O
MW: 361,01 g/mol
MDL nummer: MFCD07773044
CAS-nummer: 404001-62-3

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow Crystalline powder and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
Water 3 to 5 % (K.F.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR