5-Aminonicotinic acid 95%

Leverandør: Thermo Scientific
Warning

Synonymer: 5-Aminopyridine-3-carboxylic acid, 5-Amino-3-pyridinecarboxylic acid

361140050
ACRO361140050EA 3925 NOK
ACRO361140050
5-Aminonicotinic acid 95%
5-Aminonicotinic acid
Formel: C₆H₆N₂O₂
MW: 138,13 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00129116
CAS-nummer: 24242-19-1
FN: 2811
ADR: 6.1,II

Bestill nå

Spesifikasjon Testresultater

Appearance Golden to brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥94 %
Titration with HClO4 ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR