7-Hydroxyflavanone 98%

Leverandør: Thermo Fisher Scientific
Warning

364170010
ACRO364170010EA 540 NOK
ACRO364170010
7-Hydroxyflavanone 98%
7-Hydroxyflavanone
Formel: C₁₅H₁₂O₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00017487
CAS-nummer: 6515-36-2

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 189 to 197 °C
HPLC ≥97.5 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR