2-Iodbifenyl 98%

Leverandør: Thermo Scientific
Warning

Synonymer: 2-Iodo-1,1'-biphenyl

367020250
ACRO367020250EA 2550 NOK
ACRO367020250
2-Iodbifenyl 98%
2-Iodbifenyl
Formel: C₁₂H₉I
MW: 280,11 g/mol
Flash punkt: >110 °C
MDL nummer: MFCD00039396
CAS-nummer: 2113-51-1
EINECS: 218-303-3

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6600 to 1.6620 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR