2-Bromo-5-nitroanisole 98%

Leverandør: Thermo Fisher Scientific
Warning

370800100
ACRO370800100EA 1075 NOK
ACRO370800100
2-Bromo-5-nitroanisole 98%
2-Bromo-5-nitroanisole
Formel: C₇H₆BrNO₃
Smeltepunkt: 99…103 °C
MDL nummer: MFCD00041250
CAS-nummer: 77337-82-7
EINECS: 278-669-5

Bestill nå

Spesifikasjon Testresultater

Appearance Light brown Crystalline powder
Infrared spectrum Conforms
Melting point 99 to 103 °C
GC ≥97.5 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR