N-Boc-D-prolinal 97+%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: 1-Pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, (2R)-, tert-Butyl (R)-2-Formylpyrrolidine-1-carboxylate, (R)-N-BOC-Prolinal, (R)-2-Formylpyrrolidine-1-carboxylic acid tert-Butyl ester, N-Boc-D-Prolinal, N-Boc-D-prolinal, N-(tert-Butoxycarbonyl)-D-prolinal, (R)-tert-butyl 2-formylpyrrolidine-1-carboxylate

376230010
ACRO376230010EA 2250 NOK
ACRO376230010
N-Boc-D-prolinal 97+%
N-Boc-D-prolinal
Formel: C₁₀H₁₇NO₃
Kokepunkt: 228 °C (760 mmHg)
Tetthet: 1,059 g/cm³
Flash punkt: 110 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD01321389
CAS-nummer: 73365-02-3

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4600 to 1.4650 (20°C, 589 nm)
NMR ≥97.0 %
Specific optical rotation +90° to +102° (20°C, 589 nm) (c=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR