Benzyl-4-bromophenyl ketone

Leverandør: Thermo Fisher Scientific
Warning

381890050
ACRO381890050EA 2100 NOK
ACRO381890050
Benzyl-4-bromophenyl ketone
Benzyl-4-bromophenyl ketone
Formel: C₁₄H₁₁BrO
Smeltepunkt: 109…112 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00016331
CAS-nummer: 2001-29-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 110 to 115 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR