Methyl-3-nitrosalicylate 98%

Leverandør: Thermo Fisher Scientific
Warning

381920050
ACRO381920050EA 1700 NOK
ACRO381920050
Methyl-3-nitrosalicylate 98%
Methyl-3-nitrosalicylate
Formel: C₈H₇NO₅
Smeltepunkt: 127…132 °C
MDL nummer: MFCD00272263
CAS-nummer: 22621-41-6

Bestill nå

Spesifikasjon Testresultater

Appearance Yellow to beige to grey to brown Crystalline powder
Infrared spectrum Conforms
Melting point 127 to 132 °C
GC ≥97.5 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR