Glass Wool
Leverandør: Thermo Fisher Scientific
386060500 386060010 386062500
ACRO386060500EA 365 NOK
ACRO386060500 ACRO386060010 ACRO386062500
Glass Wool
Bomull og glassull
This glass wool can be used as a filter aid.
Boiling point: >1000 °C (760 mmHg)