Vann for analysis

Leverandør: Thermo Fisher Scientific

389400010 389400025 389400100
ACRO389400010EA 385 NOK
ACRO389400010 ACRO389400025 ACRO389400100
Vann for analysis
Vann
Formel: H₂O
Kokepunkt: 100 °C (760 mmHg)
Smeltepunkt: 0 °C
Tetthet: 1 g/cm³
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011332
CAS-nummer: 7732-18-5
EINECS: 231-791-2
Merck Indeks: 13,10098

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless Liquid
Non-volatile matter ≤0.0005 %
Residue after ign. (sulfated) ≤0.0002 %
pH 5 to 8
Absorbance ≤0.01 (water, 1 cm, 254 nm)
Chloride (Cl) ≤0.1 ppm
Nitrate (NO3) ≤0.2 ppm
Phosphate (PO4) ≤0.05 ppm
Sulfate (SO4) ≤1 ppm
Silicate (as SiO2) ≤0.01 ppm
Aluminium (Al) ≤0.02 ppm
Ammonium (NH4) ≤0.01 ppm
Antimony (Sb) ≤0.01 ppm
Barium (Ba) ≤0.01 ppm
Bismuth (Bi) ≤0.01 ppm
Boron (B) ≤0.01 ppm
Cadmium (Cd) ≤0.01 ppm
Calcium (Ca) ≤0.1 ppm
Chromium (Cr) ≤0.01 ppm
Cobalt (Co) ≤0.01 ppm
Copper (Cu) ≤0.01 ppm
Gold (Au) ≤0.01 ppm
Iron (Fe) ≤0.01 ppm
Lithium (Li) ≤0.02 ppm
Magnesium (Mg) ≤0.05 ppm
Manganese (Mn) ≤0.01 ppm
Mercury (Hg) ≤0.05 ppm
Molybdenum (Mo) ≤0.01 ppm
Nickel (Ni) ≤0.01 ppm
Potassium (K) ≤0.05 ppm
Selenium (Se) ≤0.01 ppm
Silver (Ag) ≤0.01 ppm
Sodium (Na) ≤0.1 ppm
Strontium (Sr) ≤0.05 ppm
Tin (Sn) ≤0.01 ppm
Titanium (Ti) ≤0.01 ppm
Vanadium (V) ≤0.01 ppm
Zinc (Zn) ≤0.05 ppm
Zirconium (Zr) ≤0.01 ppm
Heavy metals (as Pb) ≤0.01 ppm
KMnO4-reducing substance Passes test
Conductivity ≤6 µS/cm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR