N,N-Dimetylacetamid 99.5+% for analysis

Leverandør: Thermo Fisher Scientific
Danger

390800010
ACRO390800010EA 1200 NOK
ACRO390800010
N,N-Dimetylacetamid 99.5+% for analysis
N,N-Dimetylacetamid
Formel: CH₃CON(CH₃)₂
Kokepunkt: 164…166 °C (760 mmHg)
Smeltepunkt: –20 °C
Tetthet: 0,937 g/cm³
Flash punkt: 70 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008686
CAS-nummer: 127-19-5
EINECS: 204-826-4
ADR: 6.1,III
Merck Indeks: 13,03254
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless Liquid
Water ≤0.05 %
GC ≥99.50 %
Residue after evaporation ≤0.0005 %
Acidity ≤0.0005 meq/g
Chloride (Cl) ≤0.001 %
Sulfate (SO4) ≤0.001 %
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Nickel (Ni) ≤0.02 ppm
Zinc (Zn) ≤0.1 ppm

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR