4'-Acetamidoacetophenone 98%

Leverandør: Thermo Fisher Scientific
Warning

391820050
ACRO391820050EA 345 NOK
ACRO391820050
4'-Acetamidoacetophenone 98%
4'-Acetamidoacetophenone
Formel: C₁₀H₁₁NO₂
Smeltepunkt: 166…170 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00014965
CAS-nummer: 2719-21-3

Bestill nå

Spesifikasjon Testresultater

Appearance Beige Crystalline powder
Infrared spectrum Conforms
Melting point 166 to 170 °C
HPLC ≥97.5 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR