Trimethoxysilane 95%

Leverandør: Thermo Fisher Scientific
Danger

392170050 392170250 392171000
ACRO392170050EA 830 NOK
ACRO392170050 ACRO392170250 ACRO392171000
Trimethoxysilane 95%
Trimethoxysilane
Formel: C₃H₁₀O₃Si
Kokepunkt: 81…83 °C (1013 hPa)
Smeltepunkt: –115 °C
Tetthet: 0,96 g/cm³
Flash punkt: –9 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008340
CAS-nummer: 2487-90-3
EINECS: 219-637-2
FN: 3384
ADR: 6.1,I

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3570 to 1.3600 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR