Benzylphosphonic acid 98%

Leverandør: Thermo Fisher Scientific
Danger

393340010 393340050 393340250
ACRO393340010EA 450 NOK
ACRO393340010 ACRO393340050 ACRO393340250
Benzylphosphonic acid 98%
Benzylphosphonic acid
Formel: C₇H₉O₃P
Smeltepunkt: 165…166 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00039519
CAS-nummer: 6881-57-8
FN: 3261
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Appearance White Powder
Infrared spectrum Conforms
Melting point 169 to 173 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR