4-Methoxypiperidine 98+%

Leverandør: Thermo Fisher Scientific
Danger

393390010 393390050
ACRO393390010EA 2025 NOK
ACRO393390010 ACRO393390050
4-Methoxypiperidine 98+%
4-Methoxypiperidine
Formel: C₆H₁₄ClNO
Kokepunkt: 44 °C (9mbar)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD04115010
CAS-nummer: 4045-24-3
FN: 3267
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4590 to 1.4610 (20°C, 589 nm)
GC ≥98.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR