1-Heksansulfonsyre natriumsalt, vannfri 99+% for ioneparkromatografi

Leverandør: Thermo Fisher Scientific
Warning

396670250 396671000
ACRO396670250EA 6375 NOK
ACRO396670250 ACRO396671000
1-Heksansulfonsyre natriumsalt, vannfri 99+% for ioneparkromatografi
1-Heksansulfonsyre natriumsalt
Formel: C₆H₁₃O₃SNa
Smeltepunkt: >300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007542
CAS-nummer: 2832-45-3
EINECS: 220-601-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aebc3cb7a7cefac9ad5a7678d82a90d244b508f9&vid=d608216fe1e698d6b206b03588e6d6fe5ee661e1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline solid
Infrared spectrum Conforms
Loss on drying ≤2 % (150°C)
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.5 M soln.) Passes test
Suitability (HPLC gradient) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR