5-Methoxytryptamine 97%

Leverandør: Thermo Fisher Scientific
Danger

398580010
ACRO398580010EA 1725 NOK
ACRO398580010
5-Methoxytryptamine 97%
5-Methoxytryptamine
Formel: C₁₁H₁₄N₂O
Smeltepunkt: 120 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00005662
CAS-nummer: 608-07-1
FN: 3263
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Appearance White to tan Crystalline powder
Infrared spectrum Conforms
Melting point 118 to 123 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR