GMBS (N-Succinimidyl 4-maleimidobutyrate) 97%

Leverandør: Thermo Fisher Scientific
Warning

398681000
ACRO398681000EA 3475 NOK
ACRO398681000
GMBS (N-Succinimidyl 4-maleimidobutyrate) 97%
GMBS (N-Succinimidyl 4-maleimidobutyrate)
Formel: C₁₂H₁₂N₂O₆
Smeltepunkt: 124…132 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00036817
CAS-nummer: 80307-12-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 123 to 130 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR