Dibenzyl diisopropylphosphoramidite 90+%

Leverandør: Thermo Fisher Scientific
Warning

399640050 399640250
ACRO399640050EA 1325 NOK
ACRO399640050 ACRO399640250
Dibenzyl diisopropylphosphoramidite 90+%
Dibenzyl diisopropylphosphoramidite
Formel: C₂₀H₂₈NO₂P
Tetthet: 1,028 g/cm³
Flash punkt: 69 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00191988
CAS-nummer: 108549-23-1

Bestill nå

Spesifikasjon Testresultater

Appearance Clear to hazy colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥90.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR