Alizarin red S (sodium salt), pure, certified

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: C.I. 58005

400480250 400481000
ACRO400480250EA 1900 NOK
ACRO400480250 ACRO400481000
Alizarin red S (sodium salt), pure, certified
Alizarin red S (sodium salt)
Formel: C₁₄H₇NaO₇S
MW: 342.26 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013049
CAS-nummer: 130-22-3
EINECS: 204-981-8

Bestill nå

Spesifikasjon Testresultater

Appearance Yellow to orange to brown Powder
Infrared spectrum Conforms
UV Lambda max 590 to 599 nm
Visual transition interval from pH 9.4 (orange-brown) to pH 12.0 (violet)
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR