1,1'-Binaftyl 97%

Leverandør: Thermo Fisher Scientific
Warning

402240050
ACRO402240050EA 1575 NOK
ACRO402240050
1,1'-Binaftyl 97%
1,1'-Binaftyl
Formel: C₂₀H₁₄
Kokepunkt: 405 °C (1013 hPa)
Smeltepunkt: 143…146 °C
Tetthet: 1,19 g/cm³ (20 °C)
MDL nummer: MFCD00041740
CAS-nummer: 604-53-5
EINECS: 210-070-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to brown Powder
Infrared spectrum Conforms
GC ≥96.0 %
Titration with HClO4 ≥97.5 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR