Castor olje

Leverandør: Thermo Fisher Scientific

Synonymer: Castor oil, Ricinus oil, Oleum Ricini s. Castoris, Oil of Palma Christi

404165000 404160025
ACRO404165000EA 660 NOK
ACRO404165000 ACRO404160025
Castor olje
Castor olje
Kokepunkt: 229 °C (760 mmHg)
Smeltepunkt: –10 °C
Tetthet: 0,957 g/cm³
Flash punkt: 229 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00130746
CAS-nummer: 8001-79-4
EINECS: 232-293-8
Merck Indeks: 13,01908

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4770 to 1.4810 (20°C, 589 nm)
Acid value ≤2 mg KOH/g
Hydroxyl value ≥150 mg KOH/g
Iodine value 82 to 90 g I2/100g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR