Corn oil

Leverandør: Thermo Fisher Scientific

405430025 405435000
ACRO405430025EA 2575 NOK
ACRO405430025 ACRO405435000
Corn oil
Corn oil
Smeltepunkt: -10 °C
Tetthet: 0,92 g/cm³
Flash punkt: >200 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00130865
CAS-nummer: 8001-30-7

Bestill nå

Spesifikasjon Testresultater

Appearance Clear yellow Oily liquid
Infrared spectrum Conforms
Refractive index 1.4730 to 1.4760 (20°C, 589 nm)
Acid value ≤0.5 mg KOH/g
Peroxide value ≤5 meq O2/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR