Cyanogen iodide 98%

Leverandør: Thermo Fisher Scientific
Danger

405960050
ACRO405960050EA 2100 NOK
ACRO405960050
Cyanogen iodide 98%
Cyanogen iodide
Formel: CIN
Smeltepunkt: 142…149 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00011605
CAS-nummer: 506-78-5
EINECS: 208-053-3
FN: 1588
ADR: 6.1,II

Bestill nå

Spesifikasjon Testresultater

Appearance Off-white to light brown Crystalline powder
Infrared spectrum Conforms
Titration Iodometric ≥97.5 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR