1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) cis- and trans-mixture 99+%

Leverandør: Thermo Fisher Scientific

406040050
ACRO406040050EA 250 NOK
ACRO406040050
1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) cis- and trans-mixture 99+%
1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers)
Formel: C₈H₁₂O₄
Smeltepunkt: 165…168 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001465
CAS-nummer: 1076-97-7
EINECS: 214-068-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 164 to 168 °C
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR