Dekstran for biokjemi, high fraction

Leverandør: Thermo Fisher Scientific

406261000 406260010
ACRO406261000EA 1900 NOK
ACRO406261000 ACRO406260010
Dekstran for biokjemi, high fraction
Dekstran
Formel: (C₆H₁₀O₅)n
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00130935
CAS-nummer: 9004-54-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤9.0 %
Sulfated ash ≤2.0 %
UV at 375 nm A: ≤0.15
Viscosity 0.4 to 0.5 (1 % at 37°C)
Average molecular weight 250000
pH 4.5 to 7.0 (6 % in water)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR