Dextrin

Leverandør: Thermo Fisher Scientific

406281000 406285000
ACRO406281000EA 470 NOK
ACRO406281000 ACRO406285000
Dextrin
Dextrin
Formel: (C₆H₁₂O₆)n
Tetthet: 1,45 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00081554
CAS-nummer: 9004-53-9
EINECS: 232-675-4
Merck Indeks: 13,02971

Bestill nå

Spesifikasjon Testresultater

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Sulfated ash ≤0.5 %
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR