Erythrosin B disodium salt, pure, certified

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: C.I. 45430

409450250 409451000
ACRO409450250EA 875 NOK
ACRO409450250 ACRO409451000
Erythrosin B disodium salt, pure, certified
Erythrosin B disodium salt
Formel: C₂₀H₆I₄Na₂O₅
MW: 879.86 g/mol
Smeltepunkt: >300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00144257
CAS-nummer: 16423-68-0
EINECS: 240-474-8

Bestill nå

Spesifikasjon Testresultater

Appearance Red to brown Powder
Infrared spectrum Conforms
Dye content ≥80 %
UV Lambda max 524 to 527 nm
Absorption Ratio P-15/P+15 1.15 to 1.55
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR