4'-Ethoxyacetophenone 99%

Leverandør: Thermo Fisher Scientific
Warning

409591000 409590050 409590250 409595000
ACRO409591000EA 3350 NOK
ACRO409591000 ACRO409590050 ACRO409590250 ACRO409595000
4'-Ethoxyacetophenone 99%
4'-Ethoxyacetophenone
Formel: C₁₀H₁₂O₂
Kokepunkt: 268…269 °C (758 mmHg)
Smeltepunkt: 35…38 °C
Flash punkt: 134 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009095
CAS-nummer: 1676-63-7
EINECS: 216-825-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to yellow Crystals and/or chunks or low melting solid
Infrared spectrum Conforms
Melting point 35 to 38 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR