EDTA dinatrium salt dihydrat 99+% ACS

Leverandør: Thermo Fisher Scientific
Warning

409971000 409975000 409970025
ACRO409971000EA 680 NOK
ACRO409971000 ACRO409975000 ACRO409970025
EDTA dinatrium salt dihydrat 99+% ACS
EDTA dinatrium salt dihydrat
Formel: C₁₀H₁₄N₂Na₂O₈·2H₂O
Smeltepunkt: 252 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150037
CAS-nummer: 6381-92-6
EINECS: 205-358-3
Merck Indeks: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Titration Complexometric 99.0 to 101.0 %
pH 4.0 to 6.0 (5 % at 25°C)
Iron (Fe) ≤100 ppm
Heavy metals (as Pb) ≤50 ppm
Nitrilotriacetic acid ≤0.1 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR