Hydrogen peroksid 30% (w/w) in water Ustabilisert ACS

Leverandør: Thermo Fisher Scientific
Danger

411881000 411885000 411880010 411880025
ACRO411881000EA 420 NOK
ACRO411881000 ACRO411885000 ACRO411880010 ACRO411880025
Hydrogen peroksid 30% (w/w) in water Ustabilisert ACS
Hydrogen peroksid
Formel: H₂O₂
Kokepunkt: ∼107 °C (1013 hPa)
Smeltepunkt: –33 °C
Tetthet: 1,11 g/cm³
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00011333
CAS-nummer: 7722-84-1
EINECS: 231-765-0
FN: 2014
ADR: 5.1,II

Bestill nå

Spesifikasjon Testresultater

Appearance Clear solution
Color scale ≤10 APHA
Residue after evaporation ≤0.002 %
Titration with KMnO4 29.0 to 32.0 %
Chloride (Cl) ≤2 ppm
Nitrate (NO3) ≤2 ppm
Phosphate (PO4) ≤2 ppm
Sulfate (SO4) ≤5 ppm
Ammonium (NH4) ≤5 ppm
Iron (Fe) ≤0.5 ppm
Heavy metals (as Pb) ≤1 ppm
Titrable acid ≤0.0006 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR