Indigo carmine, ren, certified

Leverandør: Thermo Fisher Scientific
Warning

412300250 412301000
ACRO412300250EA 755 NOK
ACRO412300250 ACRO412301000
Indigo carmine, ren, certified
Indigo carmine
Formel: C₁₆H₈N₂Na₂O₈S₂
Smeltepunkt: ? 300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005723
CAS-nummer: 860-22-0
EINECS: 212-728-8
Merck Indeks: 13,04969

Bestill nå

Spesifikasjon Testresultater

Appearance Dark blue to purple Crystalline powder
Infrared spectrum Conforms
Dye content ≥80 %
UV Lambda max 608 to 612 nm
Absorption Ratio P-15/P+15 0.98 to 1.07
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR