1,2-Naphthoquinone-4-sulphonic acid sodium salt 98%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: Folin's reagent, Sodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulphonate

415400250 415401000 415400100
ACRO415400250EA 3025 NOK
ACRO415400250 ACRO415401000 ACRO415400100
1,2-Naphthoquinone-4-sulphonic acid sodium salt 98%
1,2-Naphthoquinone-4-sulphonic acid sodium salt
Formel: C₁₀H₅NaO₅S
MW: 260.20 g/mol
Smeltepunkt: 289 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001700
CAS-nummer: 521-24-4
EINECS: 208-308-9

Bestill nå

Spesifikasjon Testresultater

Appearance Orange Powder
Infrared spectrum Conforms
Titration Iodometric ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR