Nitrobenzen 99+% ACS

Leverandør: Thermo Fisher Scientific
Danger

415780250 415785000 415780025
ACRO415780250EA 665 NOK
ACRO415780250 ACRO415785000 ACRO415780025
Nitrobenzen 99+% ACS
Nitrobenzen
Formel: C₆H₅NO₂
Kokepunkt: 210,8 °C (1013 hPa)
Smeltepunkt: 5…6 °C
Tetthet: 1,205 g/cm³
Flash punkt: 88 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007043
CAS-nummer: 98-95-3
EINECS: 202-716-0
FN: 1662
ADR: 6.1,II
Merck Indeks: 13,06621

Bestill nå

Spesifikasjon Testresultater

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5500 to 1.5540 (20°C, 589 nm)
GC ≥99.0 %
Residue after evaporation ≤0.005 %
Chloride (Cl) ≤5 ppm
Water soluble titr. acid ≤0.0005 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR