L(+)-Penicillamine 98%

Leverandør: Thermo Fisher Scientific
Warning

416870010 416870050
ACRO416870010EA 750 NOK
ACRO416870010 ACRO416870050
L(+)-Penicillamine 98%
L(+)-Penicillamine
Formel: C₅H₁₁NO₂S
Smeltepunkt: 206 °C (dec.)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00064303
CAS-nummer: 1113-41-3
EINECS: 214-203-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Solubility (5 % in water) Clear colorless
Specific optical rotation +61° to +65° (20°C, 589 nm) (c=1, 1 M NaOH)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR