Pyridin 99+% ACS

Leverandør: Thermo Fisher Scientific
Danger

418545000 418540010 418540025 418540250
ACRO418545000EA 2025 NOK
ACRO418545000 ACRO418540010 ACRO418540025 ACRO418540250
Pyridin 99+% ACS
Pyridine
Formel: C₅H₅N
Kokepunkt: 115…116 °C (760 mmHg)
Smeltepunkt: –42 °C
Tetthet: 0,978 g/cm³
Flash punkt: 17 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011732
CAS-nummer: 110-86-1
EINECS: 203-809-9
FN: 1282
ADR: 3,II
Merck Indeks: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless Liquid
Water ≤0.1 % (K.F.)
Solubility (in water) Passes test
GC ≥99.0 %
Residue after evaporation ≤0.002 %
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤10 ppm
Copper (Cu) ≤5 ppm
Reducing substances Passes test
Ammonia (NH3) ≤20 ppm

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR