Resazurin sodium salt, ren, certified

Leverandør: Thermo Fisher Scientific
Warning

418900050 418900010 418900250
ACRO418900050EA 1475 NOK
ACRO418900050 ACRO418900010 ACRO418900250
Resazurin sodium salt, ren, certified
Resazurin sodium salt
Formel: C₁₂H₆NNaO₄
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005036
CAS-nummer: 62758-13-8

Bestill nå

Spesifikasjon Testresultater

Appearance Dark green to black Powder
Infrared spectrum Conforms
UV spectrum Complies
Dye content ≥80 %
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR