13-cis-Retinoic acid 99%

Leverandør: Thermo Fisher Scientific
Danger

418975000
ACRO418975000EA 4250 NOK
ACRO418975000
13-cis-Retinoic acid 99%
13-cis-Retinoic acid

Appearance: Yellow to orange Crystalline powder

Formel: C₂₀H₂₈O₂
Smeltepunkt: 174…177 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00079542
CAS-nummer: 4759-48-2

Bestill nå

Spesifikasjon Testresultater

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Melting point 174 to 177 °C
HPLC ≥98.5 %
Specific optical rotation On anhydrous basis
Titration with HClO4 97.5 to 102.5 % (On anhydrous basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR