4,4'-Bi-o-toluidin 95% pract.

Leverandør: Thermo Fisher Scientific
Danger

421120250 421121000 421122500 421125000
ACRO421120250EA 1650 NOK
ACRO421120250 ACRO421121000 ACRO421122500 ACRO421125000
4,4'-Bi-o-toluidin 95% pract.
4,4'-Bi-o-toluidin
Formel: [-C₆H₃(CH₃)-₄-NH₂]₂
Kokepunkt: ∼361 °C (1013 hPa)
Smeltepunkt: 125…132 °C
Tetthet: ~ 1,234 g/cm³ (20 °C)
Flash punkt: 244 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00014773
CAS-nummer: 119-93-7
EINECS: 204-358-0
FN: 3077
ADR: 9,III
Merck Indeks: 12,09651

Bestill nå

Spesifikasjon Testresultater

Appearance Beige to grey to brown Powder
Infrared spectrum Conforms
Melting point 125 to 132 °C
Titration with HClO4 ≥94 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR