Cupferron

Leverandør: Thermo Fisher Scientific
Danger

423601000 423605000 423600250
ACRO423601000EA 970 NOK
ACRO423601000 ACRO423605000 ACRO423600250
Cupferron
Cupferron
Formel: C₆H₉N₃O₂
Smeltepunkt: 150…155 °C (dec.)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00078422
CAS-nummer: 135-20-6
EINECS: 205-183-2
FN: 2811
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance White to yellow-brown Crystalline powder or flakes
Infrared spectrum Conforms
Additional info package usually contains small pouches of ammonium carbonate as preservative
Solubility (5 % in water) Passes test
Residue after ignition ≤0.05 %
Suitability (for precip. of Fe, etc.) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR