2,2-Dimethylcyclohexanone

Leverandør: Thermo Fisher Scientific
Warning

426500010 426500050
ACRO426500010EA 3800 NOK
ACRO426500010 ACRO426500050
2,2-Dimethylcyclohexanone
2,2-Dimethylcyclohexanone
Formel: C₈H₁₄O
Kokepunkt: 169…170 °C (,8 mmHg)
Smeltepunkt: –20 °C
Tetthet: 0,912 g/cm³
Flash punkt: 50 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00134382
CAS-nummer: 1193-47-1
FN: 1224
ADR: 3,III

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4470 to 1.4490 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR