cis-4,7,10,13,16,19-Docosahexaenoic acid 98%

Leverandør: Thermo Fisher Scientific

427640010 427641000
ACRO427640010EA 10275 NOK
ACRO427640010 ACRO427641000
cis-4,7,10,13,16,19-Docosahexaenoic acid 98%
cis-4,7,10,13,16,19-Docosahexaenoic acid
Formel: C₂₂H₃₂O₂
Smeltepunkt: –44 °C
Tetthet: 0,95 g/cm³
Flash punkt: 62 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00065722
CAS-nummer: 6217-54-5

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to light yellow Viscous liquid
Infrared spectrum Conforms
GC ≥97.5 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR