DL-α-Tocopherol (Vitamin E) (synthetic) 95%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: (±)-α-Tocopherol, Vitamin E Alcohol, all-rac-α-Tocopherol, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-ol, DL-alpha-Tocopherol, Vitamin E

428121000 428120250 428125000
ACRO428121000EA 1250 NOK
ACRO428121000 ACRO428120250 ACRO428125000
DL-α-Tocopherol (Vitamin E) (synthetic) 95%
DL-α-Tocopherol (Vitamin E)
Formel: C₂₉H₅₀O₂
MW: 430.71 g/mol
Smeltepunkt: 2.5…3.5 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00072045
CAS-nummer: 10191-41-0

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to light brown Viscous liquid
Infrared spectrum Conforms
HPLC ≥94 %
UV at 345 nm A: ≤0.022
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR