4-Chloro-3-nitropyridine 97%

Leverandør: Thermo Scientific
Danger

429930050
ACRO429930050EA 1425 NOK
ACRO429930050
4-Chloro-3-nitropyridine 97%
4-Chloro-3-nitropyridine
Formel: C₅H₃ClN₂O₂
MW: 158,54 g/mol
Smeltepunkt: 36…37,5 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD02683025
CAS-nummer: 13091-23-1
FN: 2811
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance Yellow to light brown Crystalline solid
Infrared spectrum Conforms
Melting point 35 to 39 °C
HPLC ≥96.0 %
Chloride (Cl) ≤0.0001 %
Sulfide (S) ≤0.00001 %
Phosphate (PO4) ≤0.0001 %
Carbonate (as CO2) ≤0.002 %
Aluminium (Al) ≤0.00001 %
Antimony (Sb) ≤0.000002 %
Arsenic (As) ≤0.00001 %
Barium (Ba) ≤0.00001 %
Beryllium (Be) ≤0.00001 %
Bismuth (Bi) ≤0.000005 %
Cadmium (Cd) ≤0.00001 %
Calcium (Ca) ≤0.0001 %
Chromium (Cr) ≤0.000005 %
Cobalt (Co) ≤0.00001 %
Gallium (Ga) ≤0.000005 %
Germanium (Ge) ≤0.000002 %
Gold (Au) ≤0.00001 %
Indium (In) ≤0.000005 %
Iron (Fe) ≤0.00001 %
Lead (Pb) ≤0.000005 %
Lithium (Li) ≤0.000002 %
Magnesium (Mg) ≤0.00005 %
Manganese (Mn) ≤0.000001 %
Mercury (Hg) ≤0.00001 %
Molybdenum (Mo) ≤0.000002 %
Nickel (Ni) ≤0.000005 %
Platinum (Pt) ≤0.00001 %
Potassium (K) ≤0.0001 %
Silicon (Si) ≤0.00002 %
Silver (Ag) ≤0.000002 %
Sodium (Na) ≤0.0002 %
Strontium (Sr) ≤0.0001 %
Thallium (Tl) ≤0.000002 %
Tin (Sn) ≤0.000005 %
Titanium (Ti) ≤0.000005 %
Vanadium (V) ≤0.000002 %
Zinc (Zn) ≤0.00001 %
Zirconium (Zr) ≤0.000005 %
Heavy metals (as Pb) ≤1 ppm
KMnO4-reducing substance ≤0.001 % (as O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR