Trifluoreddiksyre 99+% for HPLC

Leverandør: Thermo Fisher Scientific
Danger

432291000 432295000
ACRO432291000EA 6775 NOK
ACRO432291000 ACRO432295000
Trifluoreddiksyre 99+% for HPLC
Trifluoreddiksyre
Formel: C₂HF₃O₂
Kokepunkt: 72 °C (760 mmHg)
Smeltepunkt: –15 °C
Tetthet: 1,535 g/cm³
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004169
CAS-nummer: 76-05-1
FN: 2699
ADR: 8,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.05 %
UV at 320 nm A: ≤0.03
Color scale ≤10 APHA
Residue after evaporation ≤5 ppm
Titration with NaOH ≥99.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR