(S,S)-(+)-2-Diphenylphosphino-1,2-diphenylethylamine 97%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: (1S,2S)-2-(Diphenylphosphino)-1,2-diphenylethylamine, (1S,2S)-2-(Diphenylphosphanyl)-1,2-diphenylethan-1-amine, [(1S,2S)-1,2-Diphenyl-2-aminoethyl]diphenylphosphine, (1S,2S)-2-(Diphenylphosphino)-1,2-diphenylethanamine

434092500
ACRO434092500EA 3150 NOK
ACRO434092500
(S,S)-(+)-2-Diphenylphosphino-1,2-diphenylethylamine 97%
(S,S)-(+)-2-Diphenylphosphino-1,2-diphenylethylamine
Formel: C₂₆H₂₄NP
MW: 381.459 g/mol
Smeltepunkt: 87…93 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD11045444
CAS-nummer: 1091606-67-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
NMR ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR